Ruthenium(III) bromide (≥29% Ru)

Lieferant: Thermo Fisher Scientific
Gefahr

11807.06
11807.06EA 414 EUR
11807.06
Ruthenium(III) bromide (≥29% Ru)
Ruthenium(III) bromide

CAS No.: 14014-88-1 Molecular Formula: RuBr3 Formula Weight: 340.78 Physical Form: Powder Density: 5.30 Sensitivity: Hygroscopic Odour: Characteristic UN No.: UN3260 MDL No.: MFCD00016314

Formel: RuBr₃
Molecular Weight: 340.78 g/mol
Dichte: 5.30
Lagertemperatur: Raumtemperatur
MDL: MFCD00016314
CAS-Nummer: 14014-88-1
EINECS: 237-829-4
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR