Potassium hexacyanoruthenate(II) hydrate (≥23,0% Ru)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Potassium hexacyanoruthenate hydrate
11876.04
11876.09
11876.SS
11876.04EA
348
EUR
11876.04
11876.09
11876.SS
Potassium hexacyanoruthenate(II) hydrate (≥23,0% Ru)
Potassium hexacyanoruthenate(II) hydrate
CAS No.: 339268-21-2
Molecular Formula: C6K4N6Ru.xH2O
Formula Weight: 413.57(anhy)
Physical Form: Crystalline
Appearance: White
UN No.: UN1588
MDL No.: MFCD00168064
Formel:
C₆K₄N₆Ru·xH₂O Molecular Weight: 413.57 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168064 CAS-Nummer: 339268-21-2 EINECS: 239-097-1 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...