Potassium perruthenate(VII) ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Potassium perruthenate

11877.03 11877.06
11877.03EA 179 EUR
11877.03 11877.06
Potassium perruthenate(VII) ≥97%
Potassium perruthenate(VII)

Purity: 97% CAS No.: 10378-50-4 Molecular Formula: KO4Ru Formula Weight: 204.17 Physical Form: Crystalline Appearance: Black Melting Point: 440° dec. UN No.: UN1479 MDL No.: MFCD00061333

Formel: C₃₂H₃₆N₂O₅
Molecular Weight: 528,649 g/mol
Schmelzpunkt: 440 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00061333
CAS-Nummer: 10378-50-4
EINECS: 233-835-6
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR