Dihydrogen hexachloroosmate(IV) hydrate (≥37% Os) ≥99,95% (Metall-Basis), Premion®
Lieferant: Thermo Scientific
Gefahr
Synonyme: Chloroosmic acid, Hydrogen hexachloroosmate(IV)
12178.02 12178.03
12178.02EA 86 EUR
12178.02 12178.03
Dihydrogen hexachloroosmate(IV) hydrate (≥37% Os) ≥99,95% (Metall-Basis), Premion®
Dihydrogen hexachloroosmate(IV) hydrate
Purity: 99.95% CAS No.: 27057-71-2 Molecular Formula: H2Cl6Os.xH2O Formula Weight: 404.93(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Black Melting Point: dec. UN No.: UN3260 MDL No.: MFCD00011329
Formel: H₂Cl₆Os·xH₂O
Molecular Weight: 404,93 g/mol (anhy)
Schmelzpunkt: Zersetzung
Lagertemperatur: Raumtemperatur
MDL: MFCD00011329
CAS-Nummer: 27057-71-2
EINECS: 248-192-7
UN: 3260
ADR: 8,III