Strontium acetate hemihydrate, Reagenzqualität
Lieferant: Thermo Scientific
Synonyme:
Strontium Acetate Hemihydrate
12203.30
12203.A1
12203.30EA
74.6
EUR
12203.30
12203.A1
Strontium acetate hemihydrate, Reagenzqualität
Strontium acetate hemihydrate
Converts to SrCO₃ on ignition
Soluble in water
CAS No.: 14692-29-6
Molecular Formula: Sr(OOCCH3)2.0.5H2O
Formula Weight: 214.72 (205.71anhy)
Physical Form: Crystalline
Melting Point: 150° -0.5H2O
Solubility: Soluble in water
MDL No.: MFCD00198064
Formel:
C₈H₁₄O₉Sr₂ Molecular Weight: 429,43 g/mol Schmelzpunkt: 150 °C (-0,5H₂O) Dichte: 2,099 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00198064 CAS-Nummer: 14692-29-6 EINECS: 208-854-8 Merck Index: 14,8836 |