Iridium(III) bromide hydrate (≥35,9% Ir)

Lieferant: Thermo Fisher Scientific

12212.03 12212.06
12212.03EA 293 EUR
12212.03 12212.06
Iridium(III) bromide hydrate (≥35,9% Ir)
Iridium(III) bromide hydrate

Molecular Formula: IrBr3.xH2O Formula Weight: 431.91(anhy) Physical Form: Crystalline Melting Point: 100° -3H2O MDL No.: MFCD00049466

Formel: IrBr₃·xH₂O
Molecular Weight: 431.91 g/mol (anhy)
Schmelzpunkt: 100 °C (-3H₂O)
Lagertemperatur: Raumtemperatur
MDL: MFCD00049466
EINECS: 233-174-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR