Ruthenium(III) bromide hydrate (≥25% Ru)
Lieferant: Thermo Scientific
|
Gefahr
|
12628.03
12628.06
12628.03EA
80.9
EUR
12628.03
12628.06
Ruthenium(III) bromide hydrate (≥25% Ru)
Ruthenium(III) bromide hydrate
CAS No.: 14014-88-1
Molecular Formula: RuBr3.xH2O (x≈3)
Formula Weight: 340.78(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Odour: Characteristic
Solubility: Soluble in alcohol
UN No.: UN3260
MDL No.: MFCD00016314
Applications: Oxidation
Formel:
RuBr₃·xH₂O Molecular Weight: 340,80 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016314 CAS-Nummer: 314075-07-5 EINECS: 237-829-4 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...