Manganese(II) titanium oxide ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific

13133.09 13133.18
13133.09EA 113 EUR
13133.09 13133.18
Manganese(II) titanium oxide ≥99,9% (Metall-Basis)
Manganese(II) titanium oxide

Purity: 99.9% CAS No.: 12032-74-5 Molecular Formula: MnTiO3 Formula Weight: 150.84 Physical Form: -100 Mesh Powder Odour: Odourless Melting Point: 1360° MDL No.: MFCD00016216

Formel: MnTiO₃
Molecular Weight: 150,84 g/mol
Schmelzpunkt: 1360 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016216
CAS-Nummer: 12032-74-5
EINECS: 234-776-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR