Iron(II) titanium oxide ≥99,8% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

13137.18 13137.30
13137.18EA 172 EUR
13137.18 13137.30
Iron(II) titanium oxide ≥99,8% (Metall-Basis)
Iron(II) titanium oxide

Purity: 99.8+% CAS No.: 12022-71-8 Molecular Formula: FeTiO3 Formula Weight: 151.75 Physical Form: -100 Mesh Powder Odour: Odourless MDL No.: MFCD00064704

Formel: FeTiO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00064704
CAS-Nummer: 12022-71-8
EINECS: 234-667-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR