Neodymium(III)-2,4-pentanedionate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
Synonyme:
Neodymium(III) acetylacetonate
13184.04
13184.09
13184.04EA
72.5
EUR
13184.04
13184.09
Neodymium(III)-2,4-pentanedionate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Neodymium(III)-2,4-pentanedionate
Purity: 99.9%
CAS No.: 14589-38-9
Molecular Formula: C15H21NdO6
Formula Weight: 441.57
Physical Form: Powder
Odour: Odourless
Melting Point: 150-152°
MDL No.: MFCD00013498
Formel:
C₁₅H₂₁NdO₆ Molecular Weight: 441,57 g/mol Schmelzpunkt: 150…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013498 CAS-Nummer: 14589-38-9 EINECS: 238-631-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...