Gadolinium(III)-2,4-pentanedionate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Scientific
13204.06
13204.14
13204.06EA
86
EUR
13204.06
13204.14
Gadolinium(III)-2,4-pentanedionate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Gadolinium(III)-2,4-pentanedionate hydrate
Purity: 99.9%
CAS No.: 64438-54-6
Molecular Formula: C15H21GdO6.xH2O
Formula Weight: 454.58(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00013493
Formel:
C₁₅H₂₁GdO₆·xH₂O Molecular Weight: 454,58 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013493 CAS-Nummer: 64438-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...