Gadolinium(III) carbonate hydrate ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific

13207.14 13207.22
13207.14EA 137 EUR
13207.14 13207.22
Gadolinium(III) carbonate hydrate ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Gadolinium(III) carbonate hydrate

Purity: 99.99% CAS No.: 38245-36-2 Molecular Formula: Gd2(CO3)3.xH2O Formula Weight: 494.53(anhy) Physical Form: Powder MDL No.: MFCD00150240

Formel: Gd₂(CO₃)₃.xH₂O
Molecular Weight: 494,53 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150240
CAS-Nummer: 38245-36-2
EINECS: 227-583-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR