Europium(III) acetylacetonate hydrate ≥95% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Achtung

13209.04 13209.06 13209.09
13209.04EA 180 EUR
13209.04 13209.06 13209.09
Europium(III) acetylacetonate hydrate ≥95% (REO, Basis der Oxide der Metalle der seltenen Erden)
Europium(III) acetylacetonate hydrate

Purity: 99.9% CAS No.: 181266-82-0 Molecular Formula: C15H21EuO6.xH2O Formula Weight: 449.29(anhy) Physical Form: Powder Sensitivity: Hygroscopic Melting Point: 140° dec. MDL No.: MFCD00192170

Formel: C₁₅H₂₁EuO₆·xH₂O
Molecular Weight: 449,29 g/mol (anhy)
Schmelzpunkt: 140 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00192170
CAS-Nummer: 181266-82-0
EINECS: 238-185-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR