Yttrium(III) isopropoxide ≥90%
Lieferant: Thermo Scientific
|
Gefahr
|
14477.03
14477.06
14477.03EA
88.8
EUR
14477.03
14477.06
Yttrium(III) isopropoxide ≥90%
Yttrium(III) isopropoxide
Purity: 90+%
CAS No.: 2172-12-5
Molecular Formula: Y[OCH(CH3)2]3
Formula Weight: 266.17
Physical Form: Powder
Sensitivity: Moisture Sensitive
Melting Point: 160°
Boiling Point: 200-210°/1mm subl.
Solubility: Soluble in hexane, THF, toluene
UN No.: UN2925
MDL No.: MFCD00015641
Formel:
C₉H₂₁O₃Y Molecular Weight: 266,17 g/mol Siedepunkt: 200…210 °C (1 mmHg) (Subl.) Schmelzpunkt: 160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015641 CAS-Nummer: 2172-12-5 UN: 2925 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...