Praseodymium(III) acetate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Fisher Scientific

Synonyme: Praseodynium acetate hydrate

14611.14 14611.22
14611.14EA 77.9 EUR
14611.14 14611.22
Praseodymium(III) acetate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymium(III) acetate hydrate

Purity: 99.9% CAS No.: 6192-12-7 Molecular Formula: Pr(OOCCH3)3.xH2O Formula Weight: 318.03(anhy) Physical Form: Crystalline Aggregates MDL No.: MFCD00150121

Formel: Pr(OOCCH₃)₃·xH₂O
Molecular Weight: 318.03 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150121
CAS-Nummer: 6192-12-7
EINECS: 228-242-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR