Yttrium(III) hexafluoro-2,4-pentanedionate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Yttrium(III) hexafluoroacetylacetonate, Yttrium hexafluoroacetylacetonate

14717.03 14717.06
14717.03EA 48.5 EUR
14717.03 14717.06
Yttrium(III) hexafluoro-2,4-pentanedionate
Yttrium(III) hexafluoro-2,4-pentanedionate

CAS No.: 18911-76-7 Molecular Formula: C15H3F18O6Y Formula Weight: 710.10 Physical Form: Crystalline Melting Point: 166-170° MDL No.: MFCD00064764

Formel: C₁₅H₃F₁₈O₆Y
Schmelzpunkt: 166…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064764
CAS-Nummer: 18911-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR