Europium(III) oxalate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
|
Achtung
|
15288.04
15288.09
15288.04EA
62.7
EUR
15288.04
15288.09
Europium(III) oxalate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Europium(III) oxalate hydrate
Purity: 99.9%
CAS No.: 152864-32-9
Molecular Formula: C6Eu2O12.xH2O
Formula Weight: 567.98(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
UN No.: UN3288
MDL No.: MFCD00150143
Formel:
C₆Eu₂O₁₂·xH₂O Molecular Weight: 567,98 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150143 CAS-Nummer: 152864-32-9 EINECS: 221-885-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...