Bis-(4-bromphenyl)-ether ≥99%

Lieferant: Thermo Scientific

Synonyme: 4,4'-Dibromodiphenyl Ether, 4,4'-Oxybis(bromobenzene), 4 4'-OXYBIS(BROMOBENZENE)

16684.06 16684.14
16684.06EA 36.1 EUR
16684.06 16684.14
Bis-(4-bromphenyl)-ether ≥99%
Bis(4-bromophenyl)ether

Purity: 99% CAS No.: 2050-47-7 Molecular Formula: C12H8Br2O Formula Weight: 328.01 Physical Form: Crystalline Appearance: White Melting Point: 56-60° Boiling Point: 338-340° MDL No.: MFCD00000095

Formel: C₁₂H₈Br₂O
Molecular Weight: 328,00 g/mol
Siedepunkt: 338…340 °C
Schmelzpunkt: 56…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000095
CAS-Nummer: 2050-47-7
EINECS: 218-090-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR