Cobalt aluminate (39 - 41% Co)
Lieferant: Thermo Scientific
|
Achtung
|
17686.22
17686.36
17686.22EA
44.8
EUR
17686.22
17686.36
Cobalt aluminate (39 - 41% Co)
Cobalt aluminate
CAS No.: 12672-27-4
Molecular Formula: CoAl2O4 (approximate)
Formula Weight: 176.89
Physical Form: -200 Mesh Powder
Odour: Odourless
MDL No.: MFCD00016016
Formel:
Al₂Co₂O₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016016 CAS-Nummer: 12672-27-4 EINECS: 235-762-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...