Magnesium phosphate pentahydrate (33% P₂O₅)
Lieferant: Thermo Scientific
Achtung
Synonyme: Magnesium phosphate, tribasic pentahydrate
18219.A1 18219.A7
18219.A1EA 58.3 EUR
18219.A1 18219.A7
Magnesium phosphate pentahydrate (33% P₂O₅)
Magnesium phosphate pentahydrate
CAS No.: 10233-87-1 Molecular Formula: Mg3(PO4)2.5H2O Formula Weight: 352.93 (262.86anhy) Physical Form: Crystalline Appearance: White MDL No.: MFCD00150252
Formel: Mg₃(PO₄)₂·5H₂O
Molecular Weight: 352,93 g/mol (262,86 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150252
CAS-Nummer: 10233-87-1
EINECS: 231-824-0
Merck Index: 14,5683