Zink-Dinatrium-EDTA Hydrat, Hyrdrat

Lieferant: Thermo Fisher Scientific

Synonyme: Disodium zinc ethylenediaminetetraacetate, Ethylenediaminetetraacetic Acid Disodium Zinc Salt, Ethylenediaminetetraacetic acid zinc disodium salt

22426.14 22426.22 22426.36
22426.14EA 39.8 EUR
22426.14 22426.22 22426.36
Zink-Dinatrium-EDTA Hydrat, Hyrdrat
Ethylenediaminetetraacetic acid disodium zinc salt

CAS No.: 39208-16-7 Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2)2Zn.xH2O Formula Weight: 399.50(anhy) Physical Form: Powder MDL No.: MFCD00135187

Formel: C₁₀H₁₂N₂Na₂O₈Zn
Molecular Weight: 399.60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00135187
CAS-Nummer: 14025-21-9
EINECS: 237-865-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR