Manganese(II) methoxide
Lieferant: Thermo Scientific
|
Achtung
|
22563.03
22563.06
22563.03EA
47.4
EUR
22563.03
22563.06
Manganese(II) methoxide
Manganese(II) methoxide
CAS No.: 7245-20-7
Molecular Formula: C2H6MnO2
Formula Weight: 117.01
Physical Form: Dark powder
Sensitivity: Moisture Sensitive
Solubility: Soluble in methanol
UN No.: UN3181
MDL No.: MFCD00061476
Formel:
C₂H₆MnO₂ Molecular Weight: 117,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061476 CAS-Nummer: 7245-20-7 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...