Potassium hexacyanocobaltate(III) ≥90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tripotassium hexacyanocobaltate(III)

23126.30 23126.18
23126.30EA 323 EUR
23126.30 23126.18
Potassium hexacyanocobaltate(III) ≥90%
Potassium hexacyanocobaltate(III)

Purity: 90+% CAS No.: 13963-58-1 Molecular Formula: C6CoK3N6 Formula Weight: 332.33 Physical Form: Crystalline Density: 1.906 Appearance: Pale yellow to pale brown Odour: Odourless Melting Point: dec. Solubility: Soluble in water and acetic acid solutions. Insoluble in alcohol MDL No.: MFCD00011391

Formel: C₆CoK₃N₆
Molecular Weight: 332.332 g/mol
Schmelzpunkt: Zersetzung
Dichte: 1.906
Lagertemperatur: Raumtemperatur
MDL: MFCD00011391
CAS-Nummer: 13963-58-1
EINECS: 237-742-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR