(2,4-Pentanedionato)lithium ≥99,5%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Lithium acetylacetonate, Acetylacetone lithium salt

30529.09 30529.18 30529.30
30529.09EA 50.7 EUR
30529.09 30529.18 30529.30
(2,4-Pentanedionato)lithium ≥99,5%
(2,4-Pentanedionato)lithium

Purity: 99.5% CAS No.: 18115-70-3 Molecular Formula: C5H7LiO2 Formula Weight: 106.05 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Melting Point: ca 250°dec Beilstein Reference: 4575087 MDL No.: MFCD00013496

Formel: C₅H₇LiO₂
Molecular Weight: 106.05 g/mol
Schmelzpunkt: ∼ 250 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013496
CAS-Nummer: 18115-70-3
EINECS: 242-008-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR