Tripropylene glycol (mixture of isomers)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tripropylene glycol

31798.AK 31798.K2 31798.K7
31798.AKEA 30.1 EUR
31798.AK 31798.K2 31798.K7
Tripropylene glycol (mixture of isomers)
Tripropylene glycol (mixture of isomers)

CAS No.: 24800-44-0 Molecular Formula: HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH Formula Weight: 192.25 Physical Form: Liquid Density: 1.018 Refractive Index: 1.4443 Odour: Odourless Boiling Point: 268° MDL No.: MFCD00014405

Formel: C₉H₂₀O₄
Molecular Weight: 192.26 g/mol
Siedepunkt: 268 °C
Dichte: 1.018
Lagertemperatur: Raumtemperatur
MDL: MFCD00014405
CAS-Nummer: 24800-44-0
EINECS: 246-466-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR