Benzyl azide ≥94%

Lieferant: Thermo Scientific
Achtung

32635.04 32635.09 32635.18
32635.04EA 70.5 EUR
32635.04 32635.09 32635.18
Benzyl azide ≥94%
Benzyl azide

Purity: 94% CAS No.: 622-79-7 Molecular Formula: C6H5CH2N3 Formula Weight: 133.15 Flash Point: 43°(110°F) Physical Form: Liquid Density: 1.065 Refractive Index: 1.5341 Appearance: Colourless to pale brown Boiling Point: 82-85°/16mm Solubility: Insoluble in water UN No.: UN1993 MDL No.: MFCD00013836

Formel: C₇H₇N₃
Siedepunkt: 82…85 °C (16 mmHg)
Dichte: 1,065
Flammpunkt: 43 °C (110 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013836
CAS-Nummer: 622-79-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR