o-Carborane ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
35131.04
35131.09
35131.04EA
292
EUR
35131.04
35131.09
o-Carborane ≥98%
o-Carborane
Purity: 98%
CAS No.: 16872-09-6
Molecular Formula: C2H12B10
Formula Weight: 144.25
Physical Form: Powder
Density: 0.95
Appearance: White
Melting Point: 285-287°
Solubility: Insoluble in water
UN No.: UN1325
MDL No.: MFCD00012169
Formel:
C₂H₁₂B₁₀ Molecular Weight: 144,23 g/mol Schmelzpunkt: 285…287 °C Dichte: 0,95 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012169 CAS-Nummer: 16872-09-6 EINECS: 240-897-8 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...