Niobium-n-butoxide ≥99% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

36570.03 36570.06 36570.14
36570.03EA 62.8 EUR
36570.03 36570.06 36570.14
Niobium-n-butoxide ≥99% (Metall-Basis)
Niobium-n-butoxide

Purity: 99% CAS No.: 51030-47-8 Molecular Formula: C20H45NbO5 Formula Weight: 458.12 Physical Form: Liquid Density: 1.09 Sensitivity: Moisture Sensitive Boiling Point: 197°/0.15mm MDL No.: MFCD00144916

Formel: C₂₀H₄₅NbO₅
Molecular Weight: 458,12 g/mol
Siedepunkt: 197 °C (0,15 mmHg)
Dichte: 1,09
Lagertemperatur: Raumtemperatur
MDL: MFCD00144916
CAS-Nummer: 51030-47-8
EINECS: 256-923-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR