Neodymium(III) isopropoxide 2,5% (w/v) in 2-Propanol/ Toluol , 99% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Neodymium(III) i-propoxide

36573.AC 36573.AE
36573.ACEA 124 EUR
36573.AC 36573.AE
Neodymium(III) isopropoxide 2,5% (w/v) in 2-Propanol/ Toluol , 99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Neodymium(III) isopropoxide

Purity: 99% CAS No.: 19236-15-8 Molecular Formula: C9H21NdO3 Formula Weight: 321.30 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Blue UN No.: UN1993 MDL No.: MFCD00058819

Formel: C₉H₂₁NdO₃
Molecular Weight: 321,503 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058819
CAS-Nummer: 19236-15-8
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR