Zirconium(IV) 2-ethylhexanoate ≥97%

Lieferant: Thermo Scientific
Achtung

39174.22 39174.14
39174.22EA 219 EUR
39174.22 39174.14
Zirconium(IV) 2-ethylhexanoate ≥97%
Zirconium(IV) 2-ethylhexanoate

Purity: 97% CAS No.: 2233-42-3 Molecular Formula: C32H60O8Zr Formula Weight: 664.04 Flash Point: 93°(199°F) Physical Form: Viscous liquid Sensitivity: Moisture Sensitive Appearance: Amber MDL No.: MFCD00072684

Formel: C₃₂H₆₀O₈Zr
Molecular Weight: 664,048 g/mol
Flammpunkt: >93 °C (199 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00072684
CAS-Nummer: 2233-42-3
EINECS: 218-776-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR