Bis-[1,2-bis-(diphenylphosphino)-ethan]-palladium(0)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Pd(DIPHOS)2, Pd(dppe)2

39233.02 39233.03
39233.02EA 93.3 EUR
39233.02 39233.03
Bis-[1,2-bis-(diphenylphosphino)-ethan]-palladium(0)
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)

CAS No.: 31277-98-2 Molecular Formula: Pd[(C6H5)2PCH2CH2P(C6H5)2]2 Formula Weight: 903.25 Physical Form: Powder Sensitivity: Air Sensitive MDL No.: MFCD00009880

Formel: C₅₂H₄₈P₄Pd
Lagertemperatur: Raumtemperatur
MDL: MFCD00009880
CAS-Nummer: 31277-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR