Iron(III) hexacyanoferrate(II)
Lieferant: Thermo Fisher Scientific
Synonyme: Iron(III) Ferrocyanide, Prussian Blue
39259.14 39259.22
39259.14EA 47.3 EUR
39259.14 39259.22
Iron(III) hexacyanoferrate(II)
Iron(III) hexacyanoferrate(II)
CAS No.: 14038-43-8 Molecular Formula: C18Fe7N18 Formula Weight: 859.25 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Dark blue Odour: Odourless MDL No.: MFCD00135663
Formel: C₁₈Fe₇N₁₈
Molecular Weight: 859.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00135663
CAS-Nummer: 14038-43-8
EINECS: 237-875-5