Eisen(II)-phthalocyanin ≥96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Phthalocyanine Iron(II), Iron phthalocyanine

39262.03 39262.06
39262.03EA 42.7 EUR
39262.03 39262.06
Eisen(II)-phthalocyanin ≥96%
Iron(II) phthalocyanine

Purity: 96% CAS No.: 132-16-1 Molecular Formula: C32H16FeN8 Formula Weight: 568.38 Physical Form: Crystalline MDL No.: MFCD00015953

Formel: C₃₂H₁₆FeN₈
Molecular Weight: 568,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015953
CAS-Nummer: 132-16-1
EINECS: 205-047-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR