Dichlorobis(ethylenediamine)platinum(II) (50% Pt)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Bis(ethylenediamine)platinum(II) chloride
39280.03
39280.01
39280.03EA
250
EUR
39280.03
39280.01
Dichlorobis(ethylenediamine)platinum(II) (50% Pt)
Dichlorobis(ethylenediamine)platinum(II)
CAS No.: 21430-85-3
Molecular Formula: C4H16Cl2N4Pt
Formula Weight: 386.18
Physical Form: Crystalline
Appearance: White
Melting Point: 300° dec.
MDL No.: MFCD00009881
Formel:
C₄H₁₆Cl₂N₄Pt Molecular Weight: 386,187 g/mol Schmelzpunkt: 300 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009881 CAS-Nummer: 21430-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...