Dicarbonyl(2,4-pentanedionato)rhodium(I) 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (Acetylacetonato)dicarbonylrhodium(I), Dicarbonylacetylacetonato rhodium(I)

39295.03 39295.06 39295.MD
39295.03EA 1013 EUR
39295.03 39295.06 39295.MD
Dicarbonyl(2,4-pentanedionato)rhodium(I) 97%
Dicarbonyl(2,4-pentanedionato)rhodium(I)

Purity: 99% CAS No.: 14874-82-9 Molecular Formula: C7H7RhO4 Formula Weight: 258.04 Physical Form: Crystalline Appearance: Green Odour: Odourless Melting Point: 144-147° subl. Solubility: Soluble in acetone UN No.: UN2926 MDL No.: MFCD00009884

Formel: C₇H₈O₄Rh
Molecular Weight: 259,04 g/mol
Schmelzpunkt: 144…147 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009884
CAS-Nummer: 14874-82-9
EINECS: 238-947-9
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR