Magnesium meso-tetraphenylporphine monohydrate
Lieferant: Thermo Scientific
39316.02
39316.04
39316.02EA
127
EUR
39316.02
39316.04
Magnesium meso-tetraphenylporphine monohydrate
Magnesium meso-tetraphenylporphine monohydrate
CAS No.: 14640-21-2
Molecular Formula: C44H28MgN4.H2O
Formula Weight: 655.05 (637.04anhy)
Physical Form: Crystalline
MDL No.: MFCD00049815
Formel:
C₄₄H₂₈MgN₄·H₂O Molecular Weight: 655,05 g/mol (637,04 anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049815 CAS-Nummer: 14640-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...