Lithium periodate dihydrate, Reagenzqualität

Lieferant: Thermo Fisher Scientific
Gefahr

39355.18 39355.30
39355.18EA 64.8 EUR
39355.18 39355.30
Lithium periodate dihydrate, Reagenzqualität
Lithium periodate dihydrate

CAS No.: 21111-84-2 Molecular Formula: LiIO4.2H2O Formula Weight: 233.87 (197.84anhy) Physical Form: Powder Odour: Odourless Melting Point: 370° UN No.: UN1479 MDL No.: MFCD00049474

Formel: LiIO₄·2H₂O
Molecular Weight: 233,87 g/mol (197,84 anhy)
Schmelzpunkt: >370 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049474
CAS-Nummer: 21111-84-2
UN: 1479
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR