Magnesium niobium oxide ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific

39373.09 39373.18 39373.30
39373.09EA 83.6 EUR
39373.09 39373.18 39373.30
Magnesium niobium oxide ≥99,9% (Metall-Basis)
Magnesium niobium oxide

Purity: 99.9% CAS No.: 12163-26-7 Molecular Formula: MgNb2O6 Formula Weight: 306.11 Physical Form: -200 Mesh Powder Odour: Odourless MDL No.: MFCD00210631

Formel: MgNb₂O₆
Molecular Weight: 306,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210631
CAS-Nummer: 12163-26-7
EINECS: 235-310-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR