Carbonylchlorobis(triphenylphosphin)iridium

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Carbonylchloro bis(triphenylphosphine)iridium(I), VASKA'S COMPLEX, Chlorocarbonylbis(triphenylphosphine)iridium(I)

39413.01 39413.03 39413.06
39413.01EA 88.3 EUR
39413.01 39413.03 39413.06
Carbonylchlorobis(triphenylphosphin)iridium
Carbonylchlorobis(triphenylphosphine)iridium(I)

CAS No.: 14871-41-1 Molecular Formula: IrCl(CO)[P(C6H5)3]2 Formula Weight: 780.27 Physical Form: Crystalline Melting Point: 215° dec. Solubility: Soluble in chloroform, toluene. Slightly soluble in acetone, alcohol UN No.: UN2811 MDL No.: MFCD00009590

Formel: C₃₇H₃₀ClIrOP₂
Molecular Weight: 780.26 g/mol
Schmelzpunkt: 215 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009590
CAS-Nummer: 14871-41-1
EINECS: 238-941-6
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR