Potassium tetracyanonickelate(II) hydrate

Lieferant: Thermo Fisher Scientific
Gefahr

39434.09 39434.18
39434.09EA 103 EUR
39434.09 39434.18
Potassium tetracyanonickelate(II) hydrate
Potassium tetracyanonickelate(II) hydrate

CAS No.: 339527-86-5 Molecular Formula: K2[Ni(CN)4].xH2O Formula Weight: 240.99(anhy) Physical Form: Crystalline Density: 1.875 Melting Point: 100° -H2O Solubility: Soluble in water UN No.: UN1588 MDL No.: MFCD00049669

Formel: K₂[Ni(CN)₄].xH₂O
Molecular Weight: 240.96 g/mol (anhy)
Schmelzpunkt: 100 °C (-H₂O)
Dichte: 1.875 g/mL
Lagertemperatur: Raumtemperatur
MDL: MFCD00049669
CAS-Nummer: 339527-86-5
EINECS: 238-082-7
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR