Potassium-2-ethylhexanoate hydrate ≥95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Potassium 2-ethylhexanoate hydrate
39440.06
39440.14
39440.06EA
48.6
EUR
39440.06
39440.14
Potassium-2-ethylhexanoate hydrate ≥95%
Potassium-2-ethylhexanoate hydrate
Purity: 95%
CAS No.: 3164-85-0
Molecular Formula: C8H15KO2.xH2O
Formula Weight: 182.30(anhy)
Physical Form: Lump/semiliquid
Sensitivity: Moisture Sensitive
Odour: Odourless
MDL No.: MFCD00045896
Formel:
C₈H₁₅KO₂·xH₂O Molecular Weight: 182.30 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045896 CAS-Nummer: 3164-85-0 EINECS: 221-625-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...