Zinc meso-tetraphenylporphine
Lieferant: Thermo Fisher Scientific
Synonyme:
5,10,15,20-Tetraphenyl-21H,23H-porphine Zinc(II), Zinc(II) Tetraphenylporphyrin, 5,10,15,20-Tetraphenyl-21H,23H-porphine zinc, Zinc TPP, Zinc(II) TPP, Tetraphenylporphine zinc, TPP Zn
39552.02
39552.04
39552.02EA
129
EUR
39552.02
39552.04
Zinc meso-tetraphenylporphine
Zinc meso-tetraphenylporphine
CAS No.: 14074-80-7
Molecular Formula: C44H28N4Zn
Formula Weight: 678.10
Physical Form: Crystalline
Appearance: Purple
Odour: Odourless
MDL No.: MFCD00012155
Formel:
C₄₄H₂₈N₄Zn Molecular Weight: 678.10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012155 CAS-Nummer: 14074-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...