Bis-(ethylendithio)-tetrathiafulvalen
Lieferant: Thermo Fisher Scientific
Synonyme:
BEDT-TTF, Bis(ethylenedithiolo)tetrathiafulvalene
unter Argon abgefüllt
39557.MD
39557.03
39557.MDEA
116
EUR
39557.MD
39557.03
Bis-(ethylendithio)-tetrathiafulvalen
Bis(ethylenedithio)tetrathiafulvalene
CAS No.: 66946-48-3
Molecular Formula: C10H8S8
Formula Weight: 384.68
Physical Form: Powder- Packaged under Argon
Melting Point: 238° dec.
MDL No.: MFCD00059710
Formel:
C₁₀H₈S₈ Molecular Weight: 384.65 g/mol Schmelzpunkt: 238 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059710 CAS-Nummer: 66946-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...