Zinc hexafluoro-2,4-pentanedionate dihydrate
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Zinc hexafluoroacetylacetonate hydrate
39559.03
39559.06
39559.03EA
79.8
EUR
39559.03
39559.06
Zinc hexafluoro-2,4-pentanedionate dihydrate
Zinc hexafluoro-2,4-pentanedionate dihydrate
CAS No.: 16743-33-2
Molecular Formula: C10H2F12O4Zn.2H2O
Formula Weight: 515.49 (479.47anhy)
Physical Form: Powder
Melting Point: 157-158°
MDL No.: MFCD00150577
Formel:
C₁₀H₂F₁₂O₄Zn.2H₂O Molecular Weight: 515,541 g/mol Schmelzpunkt: 157…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150577 CAS-Nummer: 16743-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...