Silver molybdenum oxide ≥99%
Lieferant: Thermo Scientific
39650.01
39650.03
39650.01EA
65.5
EUR
39650.01
39650.03
Silver molybdenum oxide ≥99%
Silver molybdenum oxide
Purity: 99%
CAS No.: 13765-74-7
Molecular Formula: Ag2MoO4
Formula Weight: 375.68
Physical Form: Powder
Density: 6.18
Sensitivity: Light Sensitive
Appearance: Beige
Odour: Odourless
Melting Point: 483°
MDL No.: MFCD00053384
Formel:
Ag₂MoO₄ Molecular Weight: 375,68 g/mol Schmelzpunkt: 483 °C Dichte: 6,18 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053384 CAS-Nummer: 13765-74-7 EINECS: 237-374-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...