Calcium iodate monohydrate ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

40138.22 40138.36
40138.22EA 60.6 EUR
40138.22 40138.36
Calcium iodate monohydrate ≥99%
Calcium iodate monohydrate

Purity: 99+% CAS No.: 10031-32-0 Molecular Formula: Ca(IO3)2.H2O Formula Weight: 407.90 (389.88anhy) Physical Form: Powder Solubility: Slightly soluble in water UN No.: UN1479 MDL No.: MFCD00036265

Formel: Ca(IO₃)₂·H₂O
Molecular Weight: 407,90 g/mol (389,88 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036265
CAS-Nummer: 10031-32-0
EINECS: 232-191-3
UN: 1479
ADR: 5.1,II
Merck Index: 14,1676

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR