Potassium fluorosulphate ≥99,5%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
40322.09
40322.18
40322.09EA
160
EUR
40322.09
40322.18
Potassium fluorosulphate ≥99,5%
Potassium fluorosulphate
Purity: 99.5%
CAS No.: 13455-22-6
Molecular Formula: FKSO3
Formula Weight: 138.16
Physical Form: Powder/lump
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 350° dec.
UN No.: UN3260
MDL No.: MFCD00216618
Formel:
FKSO₃ Molecular Weight: 138.16 g/mol Schmelzpunkt: 350 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216618 CAS-Nummer: 13455-22-6 UN: 3260 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...