3-Hydroxy-2,2,4-trimethylpentyl isobutyrate
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate, 2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate, Isobutyric Acid 3-Hydroxy-2,2,4-trimethylpentyl Ester, 2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate)
40366.22
40366.36
40366.22EA
48.6
EUR
40366.22
40366.36
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate
CAS No.: 25265-77-4
Molecular Formula: C12H24O3
Formula Weight: 216.32
Physical Form: Liquid
Density: 0.950
Refractive Index: 1.4420
Appearance: Colourless
Melting Point: -50°
Boiling Point: 254-261°
MDL No.: MFCD00148967
Formel:
C₁₂H₂₄O₃ Molecular Weight: 216,32 g/mol Siedepunkt: 254…261 °C Schmelzpunkt: –50 °C Dichte: 0,950 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148967 CAS-Nummer: 25265-77-4 EINECS: 246-771-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...