Potassium hexachlororuthenate(IV) (metal basis) (≥25,3% Ru) ≥99,95%, Premion®

Lieferant: Thermo Scientific
Achtung

40399.03 40399.06
40399.03EA 79.1 EUR
40399.03 40399.06
Potassium hexachlororuthenate(IV) (metal basis) (≥25,3% Ru) ≥99,95%, Premion®
Potassium hexachlororuthenate(IV) (metal basis)

Purity: 99.95% CAS No.: 23013-82-3 Molecular Formula: K2RuCl6 Formula Weight: 391.99 Physical Form: Cystalline Powder MDL No.: MFCD00050159

Formel: K₂RuCl₆
Molecular Weight: 391,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050159
CAS-Nummer: 23013-82-3
EINECS: 245-381-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR