Hydrogen tetrabromoaurate(III) hydrate (≥32% Au) ≥99,99% (Metall-Basis), Premion®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Bromoauric acid
40433.03
40433.06
40433.03EA
131
EUR
40433.03
40433.06
Hydrogen tetrabromoaurate(III) hydrate (≥32% Au) ≥99,99% (Metall-Basis), Premion®
Hydrogen tetrabromoaurate(III) hydrate
Purity: 99.99%
Molecular Formula: HAuBr4.xH2O (x≈5)
Formula Weight: 517.61(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: ca 27°
Solubility: Soluble in water and alcohol
UN No.: UN3260
MDL No.: MFCD00054118
Formel:
HAuBr₄·xH₂O(x≈3) Molecular Weight: 517,61 g/mol (anhy) Schmelzpunkt: ∼ 27 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054118 CAS-Nummer: 17083-68-0 UN: 3260 ADR: 8,III |